siRNA Details
| Gene Symbol: Spdyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015385.1 | - | 1, 2 | 267 | ||
| BC141200.1 | - | 1, 2 | 267 | 9055812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spdyb| Gene Symbol: Spdyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015385.1 | - | 1, 2 | 267 | ||
| BC141200.1 | - | 1, 2 | 267 | 9055812 | ||