siRNA Details
| Gene Symbol: Spata24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027733.5 | NP_082009.3 | 1 | 138 | ||
| NM_029485.2 | NP_083761.1 | 1 | 138 | |||
| GenBank mRNA | AK012241.1 | - | 1 | 110 | ||
| AK016341.1 | - | 1 | 110 | |||
| AK017218.1 | - | 1 | 114 | |||
| AY730532.1 | - | 1 | 138 | |||
| BC048524.1 | - | 1 | 87 | |||
| BC060950.1 | - | 1 | 91 | 6773733 | ||
| BU961109.1 | - | 1 | 141 | |||

Spata24
Genomic Map