siRNA Details
| Gene Symbol: Vmn1r172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030735.1 | NP_109660.1 | 2 | 1215 | ||
| GenBank mRNA | AF324873.1 | - | 2 | 1215 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r172| Gene Symbol: Vmn1r172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030735.1 | NP_109660.1 | 2 | 1215 | ||
| GenBank mRNA | AF324873.1 | - | 2 | 1215 | ||