siRNA Details
| Gene Symbol: Ly6g6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033478.2 | NP_258439.1 | 3 | 528 | ||
| GenBank mRNA | BB737333.1 | - | 1 | 324 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6g6d| Gene Symbol: Ly6g6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033478.2 | NP_258439.1 | 3 | 528 | ||
| GenBank mRNA | BB737333.1 | - | 1 | 324 | ||