siRNA Details
| Gene Symbol: Vmn1r16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134184.2 | NP_598945.2 | 1 | 853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r16| Gene Symbol: Vmn1r16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134184.2 | NP_598945.2 | 1 | 853 | ||