siRNA Details
| Gene Symbol: Obox7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038676.1 | NP_001033765.1 | 4 | 874 | ||
| GenBank mRNA | BC099488.1 | - | 4 | 874 | 30939375 | |
| Gene Symbol: Obox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145709.2 | NP_663755.2 | 4 | 906 | ||
| GenBank mRNA | BB701159.1 | - | 1 | 75 | ||
| DV653414.1 | - | 2 | 471 | |||

Obox7
Genomic Map