siRNA Details
| Gene Symbol: Olfr1511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146271.2 | NP_666383.2 | 1 | 289 | ||
| GenBank mRNA | BC112968.1 | - | 1 | 287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1511| Gene Symbol: Olfr1511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146271.2 | NP_666383.2 | 1 | 289 | ||
| GenBank mRNA | BC112968.1 | - | 1 | 287 | ||