siRNA Details
| Gene Symbol: Olfr725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146317.2 | NP_666429.2 | 1 | 877 | ||
| GenBank mRNA | BC125411.1 | - | 1 | 877 | 40129926 | |
| BC125417.1 | - | 1 | 877 | 40129932 | ||
| Gene Symbol: Olfr726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146316.2 | NP_666428.2 | 1 | 778 | ||

Olfr725
Genomic Map