siRNA Details
| Gene Symbol: MRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317947.1 | NP_001304876.1 | 1 | 262 | ||
| NM_018146.3 | NP_060616.1 | 1 | 262 | |||
| GenBank mRNA | AF177344.2 | - | 1 | 318 | ||
| AK001443.1 | - | 1 | 238 | |||
| AK222900.1 | - | 1 | 238 | |||
| AK225128.1 | - | 1 | 219 | |||
| BC011550.1 | - | 1 | 224 | 4620063 | ||
| BC050614.1 | - | 1 | 220 | 5248052 | ||
| BI830018.1 | - | 1 | 223 | |||

MRM3
Genomic Map