siRNA Details
| Gene Symbol: Olfr190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104050.1 | - | 1 | 46 | 40040225 | |
| BC104051.1 | - | 1 | 46 | 40040226 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr190| Gene Symbol: Olfr190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104050.1 | - | 1 | 46 | 40040225 | |
| BC104051.1 | - | 1 | 46 | 40040226 | ||