siRNA Details
| Gene Symbol: Olfr875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146749.2 | NP_666960.2 | 1 | 495 | ||
| GenBank mRNA | BC132237.1 | - | 1 | 495 | 40130514 | |
| BC138017.1 | - | 1 | 495 | 8861036 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr875| Gene Symbol: Olfr875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146749.2 | NP_666960.2 | 1 | 495 | ||
| GenBank mRNA | BC132237.1 | - | 1 | 495 | 40130514 | |
| BC138017.1 | - | 1 | 495 | 8861036 | ||