siRNA Details
Gene Symbol: Olfr875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146749.2 | NP_666960.2 | 1 | 286 | ||
GenBank mRNA | BC132237.1 | - | 1 | 286 | 40130514 | |
BC138017.1 | - | 1 | 286 | 8861036 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146749.2 | NP_666960.2 | 1 | 286 | ||
GenBank mRNA | BC132237.1 | - | 1 | 286 | 40130514 | |
BC138017.1 | - | 1 | 286 | 8861036 |