siRNA Details
| Gene Symbol: Olfr1502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146797.1 | NP_667008.1 | 1 | 917 | ||
| GenBank mRNA | BC106809.2 | - | 760 | |||
| BC106810.1 | - | 1 | 917 | 40042985 | ||
| BC106813.2 | - | 760 | ||||
| BC106814.2 | - | 760 | ||||

Olfr1502
Genomic Map