siRNA Details
| Gene Symbol: Olfr1502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146797.1 | NP_667008.1 | 1 | 274 | ||
| GenBank mRNA | BC106810.1 | - | 1 | 274 | 40042985 | |
| Gene Symbol: Olfr1505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC150832.1 | - | 1 | 354 | 9087743 | |

Olfr1502
Genomic Map