siRNA Details
| Gene Symbol: Olfr101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146834.1 | NP_667045.1 | 1 | 812 | ||
| GenBank mRNA | BC138322.1 | - | 1 | 876 | 8861342 | |
| BC138323.1 | - | 1 | 876 | 8861343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr101| Gene Symbol: Olfr101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146834.1 | NP_667045.1 | 1 | 812 | ||
| GenBank mRNA | BC138322.1 | - | 1 | 876 | 8861342 | |
| BC138323.1 | - | 1 | 876 | 8861343 | ||