siRNA Details
| Gene Symbol: Olfr763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146862.1 | NP_667073.1 | 1 | 821 | ||
| GenBank mRNA | BC119229.1 | - | 1 | 886 | 8733978 | |
| BC119231.1 | - | 1 | 886 | 8733980 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr763| Gene Symbol: Olfr763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146862.1 | NP_667073.1 | 1 | 821 | ||
| GenBank mRNA | BC119229.1 | - | 1 | 886 | 8733978 | |
| BC119231.1 | - | 1 | 886 | 8733980 | ||