siRNA Details
| Gene Symbol: Olfr330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146879.2 | NP_667090.1 | 2 | 802 | ||
| GenBank mRNA | BC120838.1 | - | 1 | 862 | 40129761 | |
| BC172163.1 | - | 1 | 780 | |||
| BC172164.1 | - | 1 | 780 | |||
| BC172166.1 | - | 1 | 780 | |||

Olfr330
Genomic Map