siRNA Details
Gene Symbol: Olfr1228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146971.1 | NP_667182.1 | 1 | 352 | ||
GenBank mRNA | BC125451.1 | - | 1 | 435 | 40129966 | |
BC125453.1 | - | 1 | 435 | 40129968 |
