siRNA Details
| Gene Symbol: Olfr181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146999.2 | NP_667210.2 | 2 | 391 | ||
| GenBank mRNA | BC125397.1 | - | 1 | 104 | 40129912 | |
| BC131916.1 | - | 1 | 104 | 40130193 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB172861.1 | - | 348 | Chr. - - [-] on Build | ||

Olfr181
Genomic Map