siRNA Details
| Gene Symbol: Olfr584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147054.1 | NP_667265.1 | 1 | 454 | ||
| GenBank mRNA | BC128007.1 | - | 1 | 458 | 40135987 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr584| Gene Symbol: Olfr584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147054.1 | NP_667265.1 | 1 | 454 | ||
| GenBank mRNA | BC128007.1 | - | 1 | 458 | 40135987 | |