siRNA Details
| Gene Symbol: EVA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304762.1 | NP_001291691.1 | 3 | 952 | ||
| NM_018166.2 | NP_060636.1 | 3 | 1024 | |||
| NR_130899.1 | 2 | 855 | ||||
| XM_017001627.1 | XP_016857116.1 | 2 | 1146 | |||
| GenBank mRNA | AK001509.1 | - | 3 | 1024 | ||
| BC006241.2 | - | 2 | 736 | 3951625 | ||
| BC071697.1 | - | 2 | 943 | 4371667 | ||

EVA1B
Genomic Map