siRNA Details
| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014283.1 | NP_001014305.1 | 2, 3 | 259 | ||
| XM_017020643.1 | XP_016876132.1 | 2, 3 | 470 | |||
| XM_017020644.1 | XP_016876133.1 | 2, 3 | 400 | |||
| XM_017020645.1 | XP_016876134.1 | 2, 3 | 441 | |||
| XM_017020646.1 | XP_016876135.1 | 2, 3 | 4182 | |||
| XM_017020647.1 | XP_016876136.1 | 2, 3 | 720 | |||
| XM_017020648.1 | XP_016876137.1 | 2, 3 | 1483 | |||
| GenBank mRNA | AK001566.1 | - | 2, 3 | 242 | ||
| BC056669.1 | - | 2, 3 | 246 | 6494926 | ||
| BM836766.1 | - | 2, 3 | 402 | |||

DCUN1D2
Genomic Map