siRNA Details
| Gene Symbol: DPPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018189.3 | NP_060659.3 | 2 | 190 | ||
| XM_011512954.2 | XP_011511256.1 | 2 | 469 | |||
| GenBank mRNA | AK001575.1 | - | 2 | 132 | ||
| AK022698.1 | - | 2 | 147 | |||
| AK290992.1 | - | 2 | 183 | |||
| AK298623.1 | - | 2 | 140 | |||
| AK299290.1 | - | 2 | 133 | |||
| BC032846.2 | - | 2 | 164 | 5267235 | ||

DPPA4
Genomic Map