siRNA Details
| Gene Symbol: DNAJC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018198.3 | NP_060668.2 | 2 | 221 | ||
| XM_017001745.1 | XP_016857234.1 | 2 | 148 | |||
| GenBank mRNA | AF306694.1 | - | 2 | 120 | ||
| AF306695.1 | - | 2 | 135 | |||
| AK001599.1 | - | 2 | 130 | |||
| AK294539.1 | - | 2 | 118 | |||
| AK298378.1 | - | 2 | 107 | |||
| AL109978.1 | - | 1 | 43 | |||
| AL833841.1 | - | 2 | 95 | |||
| AY336749.1 | - | 2 | 131 | |||
| BC006086.1 | - | 2 | 117 | 3946414 | ||
| BC008772.1 | - | 2 | 122 | |||
| BC014145.2 | - | 2 | 129 | 4762069 | ||
| DB461237.1 | - | 2 | 223 | |||

DNAJC11
Genomic Map