siRNA Details
| Gene Symbol: KIAA0087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_022006.1 | 2 | 731 | |||
| GenBank mRNA | BC128257.1 | - | 2 | 524 | ||
| D42038.1 | - | 2 | 694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA0087| Gene Symbol: KIAA0087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_022006.1 | 2 | 731 | |||
| GenBank mRNA | BC128257.1 | - | 2 | 524 | ||
| D42038.1 | - | 2 | 694 | |||