siRNA Details
| Gene Symbol: SNHG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024127.1 | 2 | 349 | |||
| GenBank mRNA | AF277181.1 | - | 2 | 137 | ||
| AK092096.1 | - | 1 | 730 | |||
| BC022205.1 | - | 2 | 978 | |||
| BC034611.1 | - | 2 | 159 | |||
| BC045792.1 | - | 2 | 367 | |||
| BC064589.1 | - | 2 | 337 | |||
| BC068048.1 | - | 2 | 349 | |||
| KJ782215.1 | - | 3 | 1478 | |||

SNHG12
Genomic Map