siRNA Details
| Gene Symbol: ADGRL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022159.3 | NP_071442.2 | 2 | 285 | ||
| GenBank mRNA | AB032017.1 | - | 2 | 246 | ||
| AK292894.1 | - | 2 | 178 | |||
| AK313738.1 | - | 2 | 284 | |||
| AY358360.1 | - | 2 | 152 | |||
| BC025721.1 | - | 2 | 151 | 5229055 | ||
| DA477962.1 | - | 2 | 286 | |||
More Information
Set Membership: |
|

ADGRL4
Genomic Map