siRNA Details
| Gene Symbol: C10orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039380.3 | NP_001034469.2 | 2 | 1439 | ||
| NR_126421.1 | 3 | 576 | ||||
| XM_017015895.1 | XP_016871384.1 | 3 | 570 | |||
| GenBank mRNA | AK055129.1 | - | 2 | 1417 | ||

C10orf25
Genomic Map