siRNA Details
| Gene Symbol: CRYBG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153605.3 | NP_705833.3 | 4 | 1758 | ||
| XM_005247117.4 | XP_005247174.1 | 3 | 1461 | |||
| GenBank mRNA | AK296966.1 | - | 4 | 1758 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYBG3| Gene Symbol: CRYBG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153605.3 | NP_705833.3 | 4 | 1758 | ||
| XM_005247117.4 | XP_005247174.1 | 3 | 1461 | |||
| GenBank mRNA | AK296966.1 | - | 4 | 1758 | ||