siRNA Details
| Gene Symbol: PNMAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018215.3 | NP_060685.2 | 2 | 1504 | ||
| XM_011527067.2 | XP_011525369.1 | 2 | 1518 | |||
| GenBank mRNA | AK001643.1 | - | 1 | 523 | ||
| AK096326.1 | - | 2 | 1064 | |||
| AK290218.1 | - | 3 | 1387 | |||
| BC026026.1 | - | 1 | 537 | |||
| BC032508.1 | - | 2 | 1482 | 5246377 | ||
| BC051688.1 | - | 2 | 1461 | 5295669 | ||

PNMAL1
Genomic Map