siRNA Details
| Gene Symbol: WDR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291088.1 | NP_001278017.1 | 6 | 6764 | ||
| NM_031951.4 | NP_114157.4 | 6 | 6647 | |||
| XM_011527360.2 | XP_011525662.1 | 6 | 6591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR87| Gene Symbol: WDR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291088.1 | NP_001278017.1 | 6 | 6764 | ||
| NM_031951.4 | NP_114157.4 | 6 | 6647 | |||
| XM_011527360.2 | XP_011525662.1 | 6 | 6591 | |||