siRNA Details
| Gene Symbol: CSN1S2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003720.1 | 11 | 442 | |||
| GenBank mRNA | AY154892.1 | - | 11 | 442 | ||
| BC128170.1 | - | 8 | 316 | |||
| BC128173.1 | - | 8 | 316 | |||
| BC141914.1 | - | 8 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSN1S2AP| Gene Symbol: CSN1S2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003720.1 | 11 | 442 | |||
| GenBank mRNA | AY154892.1 | - | 11 | 442 | ||
| BC128170.1 | - | 8 | 316 | |||
| BC128173.1 | - | 8 | 316 | |||
| BC141914.1 | - | 8 | 316 | |||