siRNA Details
| Gene Symbol: VWA3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173615.4 | NP_775886.3 | 24 | 2540 | ||
| XM_011545742.2 | XP_011544044.1 | 27 | 2750 | |||
| XM_011545744.2 | XP_011544046.1 | 12 | 1350 | |||
| XM_011545745.1 | XP_011544047.1 | 10 | 1118 | |||
| XM_011545746.2 | XP_011544048.1 | 10 | 1221 | |||
| XM_017022947.1 | XP_016878436.1 | 28 | 2823 | |||
| XM_017022948.1 | XP_016878437.1 | 11 | 1922 | |||
| XM_017022949.1 | XP_016878438.1 | 9 | 2193 | |||
| GenBank mRNA | AK128606.1 | - | 13 | 1408 | ||
| BC109294.1 | - | 13 | 1315 | |||
| BC109295.1 | - | 12 | 1268 | |||

VWA3A
Genomic Map