siRNA Details
| Gene Symbol: VWA3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173615.4 | NP_775886.3 | 34 | 3889 | ||
| XM_011545742.2 | XP_011544044.1 | 38 | 4171 | |||
| XM_011545744.2 | XP_011544046.1 | 23 | 2771 | |||
| XM_011545745.1 | XP_011544047.1 | 21 | 2539 | |||
| XM_011545746.2 | XP_011544048.1 | 21 | 2642 | |||
| XM_017022947.1 | XP_016878436.1 | 39 | 4244 | |||
| XM_017022948.1 | XP_016878437.1 | 22 | 3343 | |||
| XM_017022949.1 | XP_016878438.1 | 20 | 3614 | |||
| GenBank mRNA | AK093467.1 | - | 10 | 1452 | ||
| AK128606.1 | - | 23 | 2757 | |||
| BC038400.1 | - | 11 | 1334 | |||

VWA3A
Genomic Map