siRNA Details
| Gene Symbol: RNPEPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018226.4 | NP_060696.4 | 4, 5 | 946 | ||
| XM_005247036.4 | XP_005247093.1 | 4, 5 | 1007 | |||
| GenBank mRNA | AF300795.1 | - | 4, 5 | 652 | ||
| AK130805.1 | - | 1, 2 | 30 | |||
| BC017301.2 | - | 1, 2 | 98 | |||
| BC067258.1 | - | 4, 5 | 521 | 6183046 | ||
| BC082975.1 | - | 4, 5 | 846 | 6379537 | ||
| BC099906.1 | - | 4 | 893 | 5311883 | ||

RNPEPL1
Genomic Map