siRNA Details
| Gene Symbol: MAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019090.2 | NP_061963.2 | 1 | 345 | ||
| GenBank mRNA | BC109122.2 | - | 1 | 211 | 40036000 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP10| Gene Symbol: MAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019090.2 | NP_061963.2 | 1 | 345 | ||
| GenBank mRNA | BC109122.2 | - | 1 | 211 | 40036000 | |