siRNA Details
| Gene Symbol: LINC00282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027047.1 | 5 | 2714 | |||
| NR_027048.1 | 3 | 2571 | ||||
| GenBank mRNA | AK094626.1 | - | 1 | 1260 | ||
| AK124707.1 | - | 3 | 2571 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00282| Gene Symbol: LINC00282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027047.1 | 5 | 2714 | |||
| NR_027048.1 | 3 | 2571 | ||||
| GenBank mRNA | AK094626.1 | - | 1 | 1260 | ||
| AK124707.1 | - | 3 | 2571 | |||