siRNA Details
| Gene Symbol: ERICH6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182542.2 | NP_872348.2 | 15 | 2336 | ||
| XM_011534965.2 | XP_011533267.1 | 14 | 2355 | |||
| XM_017020418.1 | XP_016875907.1 | 14 | 2283 | |||
| GenBank mRNA | AK057244.1 | - | 15 | 2318 | ||

ERICH6B
Genomic Map