siRNA Details
| Gene Symbol: LINC00663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026956.1 | 3 | 2237 | |||
| GenBank mRNA | AK022418.1 | - | 1 | 1873 | ||
| BC034236.1 | - | 5 | 1954 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00663| Gene Symbol: LINC00663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026956.1 | 3 | 2237 | |||
| GenBank mRNA | AK022418.1 | - | 1 | 1873 | ||
| BC034236.1 | - | 5 | 1954 | |||