siRNA Details
Gene Symbol: ATAD3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039211.2 | NP_001034300.2 | 11 | 2016 | ||
GenBank mRNA | AK091918.1 | - | 11 | 2017 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ATAD3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039211.2 | NP_001034300.2 | 11 | 2016 | ||
GenBank mRNA | AK091918.1 | - | 11 | 2017 |