siRNA Details
| Gene Symbol: CCDC137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199287.2 | NP_954981.1 | 1, 2 | 178 | ||
| XM_011524738.2 | XP_011523040.1 | 1, 2 | 85 | |||
| XM_017024573.1 | XP_016880062.1 | 1, 2 | 212 | |||
| GenBank mRNA | BC009369.2 | - | 1, 2 | 162 | 4110481 | |
| BM548065.1 | - | 1, 2 | 217 | |||
| FJ649645.1 | - | 1, 2 | 1868 | |||

CCDC137
Genomic Map