siRNA Details
| Gene Symbol: ILF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024333.1 | 1 | 1928 | |||
| GenBank mRNA | AF439744.2 | - | 1 | 1538 | ||
| AL832185.1 | - | 1 | 1902 | |||
| BC044911.2 | - | 1 | 1904 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ILF3-AS1| Gene Symbol: ILF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024333.1 | 1 | 1928 | |||
| GenBank mRNA | AF439744.2 | - | 1 | 1538 | ||
| AL832185.1 | - | 1 | 1902 | |||
| BC044911.2 | - | 1 | 1904 | |||