siRNA Details
| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099652.1 | NP_001093122.1 | 7 | 3132 | ||
| GenBank mRNA | BM842962.1 | - | 1 | 224 | ||
| BX647179.1 | - | 5 | 4653 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR137C| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099652.1 | NP_001093122.1 | 7 | 3132 | ||
| GenBank mRNA | BM842962.1 | - | 1 | 224 | ||
| BX647179.1 | - | 5 | 4653 | |||