siRNA Details
Gene Symbol: TAS1R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152228.2 | NP_689414.1 | 2 | 251 | ||
XM_017002435.1 | XP_016857924.1 | 2 | 273 | |||
XM_017002436.1 | XP_016857925.1 | 2 | 273 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
