siRNA Details
Gene Symbol: OC90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080399.2 | NP_001073868.2 | 11, 12 | 938 | ||
GenBank mRNA | AK297911.1 | - | 12, 13 | 938 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OC90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080399.2 | NP_001073868.2 | 11, 12 | 938 | ||
GenBank mRNA | AK297911.1 | - | 12, 13 | 938 |