siRNA Details
| Gene Symbol: FAM171A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198475.2 | NP_940877.2 | 3 | 553 | ||
| GenBank mRNA | BC029298.1 | - | 3 | 569 | ||
| BC030200.1 | - | 3 | 553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM171A2| Gene Symbol: FAM171A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198475.2 | NP_940877.2 | 3 | 553 | ||
| GenBank mRNA | BC029298.1 | - | 3 | 569 | ||
| BC030200.1 | - | 3 | 553 | |||