siRNA Details
Gene Symbol: FAM171A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198475.2 | NP_940877.2 | 2, 3 | 500 | ||
GenBank mRNA | BC029298.1 | - | 2, 3 | 516 | ||
BC030200.1 | - | 2, 3 | 500 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM171A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198475.2 | NP_940877.2 | 2, 3 | 500 | ||
GenBank mRNA | BC029298.1 | - | 2, 3 | 516 | ||
BC030200.1 | - | 2, 3 | 500 |