siRNA Details
| Gene Symbol: EMC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103821.1 | 3 | 1307 | |||
| GenBank mRNA | AK092352.1 | - | 2 | 1255 | ||
| AK125558.1 | - | 4 | 1076 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMC3-AS1| Gene Symbol: EMC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103821.1 | 3 | 1307 | |||
| GenBank mRNA | AK092352.1 | - | 2 | 1255 | ||
| AK125558.1 | - | 4 | 1076 | |||