siRNA Details
| Gene Symbol: PSMG3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021487.2 | 6 | 3343 | |||
| NR_027328.1 | 4 | 5184 | ||||
| NR_027329.1 | 5 | 5292 | ||||
| GenBank mRNA | AB067495.1 | - | 4 | 5162 | ||
| AK091044.1 | - | 1 | 2539 | |||
| AK092212.1 | - | 1 | 1786 | |||
| BC036405.1 | - | 6 | 3151 | |||
| BC047649.1 | - | 6 | 3146 | |||

PSMG3-AS1
Genomic Map