siRNA Details
| Gene Symbol: AGAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133447.1 | NP_597704.1 | 12 | 1824 | ||
| GenBank mRNA | AB075855.1 | - | 7 | 3228 | ||
| AK097165.1 | - | 12 | 1824 | |||
| BC140762.1 | - | 8 | 1070 | 9021630 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGAP11| Gene Symbol: AGAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133447.1 | NP_597704.1 | 12 | 1824 | ||
| GenBank mRNA | AB075855.1 | - | 7 | 3228 | ||
| AK097165.1 | - | 12 | 1824 | |||
| BC140762.1 | - | 8 | 1070 | 9021630 | ||